AC-Tyr-NHME

Supplier: BACHEM AMERICAS INC

4003924.0250 4003924.1000
E-1255.0250BAEA 211.77 CAD
E-1255.0250BA E-1255.1000BA
AC-Tyr-NHME
AC-Tyr-NHME

Sequence: Ac-Tyr-NHMe


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₂H₁₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00237518
CAS Number: 6367-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR