Ac-Tyr-NH₂

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Acetamido-3-(4-hydroxyphenyl)propanamide, N-Acetyl-L-tyrosinamide

4001968.0001 4001968.0005
E-1245.0001BAEA 180.91 CAD
E-1245.0001BA E-1245.0005BA
Ac-Tyr-NH₂
Ac-Tyr-NH₂

Sequence: Ac-Tyr-NH₂


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₁H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00002391
CAS Number: 1948-71-6
EINECS: 217-756-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR