N-Acetyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

4026352.0025 4026352.0100 4026352.0250
E-1240.0025BAEA 180.91 CAD
E-1240.0025BA E-1240.0100BA E-1240.0250BA
N-Acetyl-L-tyrosine
N-Acetyl-L-tyrosine

Sequence: Ac-Tyr-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.


Synonym(s): NAT

Formula: C₁₁H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037190
CAS Number: 537-55-3
EINECS: 208-671-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR