Ac-Trp-NH₂

Supplier: BACHEM AMERICAS INC

4002401.0001 4002401.0005
E-1225.0001BAEA 187.69 CAD
E-1225.0001BA E-1225.0005BA
Ac-Trp-NH₂
Ac-Trp-NH₂

Sequence: Ac-Trp-NH₂
Synonym(s): NATA


Reference compound for the determination of quantum yields in proteins.

Formula: C₁₃H₁₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00005646
CAS Number: 2382-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR