(S)-Methyl-2-acetamido-3-hydroxypropanoate

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Ser-OMe

4000152.0250 4000152.1000
E-1205.0250BAEA 180.91 CAD
E-1205.0250BA E-1205.1000BA
(S)-Methyl-2-acetamido-3-hydroxypropanoate
(S)-Methyl-2-acetamido-3-hydroxypropanoate

Sequence: Ac-Ser-OMe


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₆H₁₁NO₄
MW: 161.157 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038508
CAS Number: 54322-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR