O-Acetyl-L-serine hydrochloride

Supplier: BACHEM AMERICAS INC

4001221.0001 4001221.0005
E-1200.0001BAEA 185.3 CAD
E-1200.0001BA E-1200.0005BA
O-Acetyl-L-serine hydrochloride
O-Acetyl-L-serine hydrochloride

Sequence: H-Ser(Ac)-OH · HCl


Bachem also offers derivatives of phosphoserine and sulfoserine.


O-Acetylserine is a substrate for cysteine synthase and β-substituted alanine synthase in higher plants and appears to be a naturally occurring biosynthetic key intermediate.

Formula: C₅H₉NO₄
Storage Temperature: Freezer
MDL Number: MFCD00060169
CAS Number: 66638-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR