N-Acetyl-L-prolinamide

Supplier: BACHEM AMERICAS INC

4007095.0001 4007095.0005
E-1185.0001BAEA 180.91 CAD
E-1185.0001BA E-1185.0005BA
N-Acetyl-L-prolinamide
N-Acetyl-L-prolinamide

Sequence: Ac-Pro-NH₂

Formula: C₇H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00037340
CAS Number: 16395-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR