N-Acetyl-L-proline
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Acetyl-L-Proline, Ac-Pro-OH
4002573.0005
4002573.0025
E-1180.0005BAEA
180.91
CAD
E-1180.0005BA
E-1180.0025BA
N-Acetyl-L-proline
N-Acetyl-L-proline
Sequence: Ac-Pro-OH
Formula:
C₇H₁₁NO₃ MW: 157.169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00020837 CAS Number: 68-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...