N-Acetylornithine
Supplier: BACHEM AMERICAS INC
4002042.0001
4002042.0005
E-1150.0001BAEA
206.51
CAD
E-1150.0001BA
E-1150.0005BA
N-Acetylornithine
N-Acetylornithine
Sequence: Ac-Orn-OH
Formula:
C₇H₁₄N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00065115 CAS Number: 6205-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...