N-Acetyl-L-methionine

Supplier: BACHEM AMERICAS INC

4003668.0005 4003668.0025
E-1135.0005BAEA 180.91 CAD
E-1135.0005BA E-1135.0025BA
N-Acetyl-L-methionine
N-Acetyl-L-methionine

Sequence: Ac-Met-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.


Competitive inhibitor of acyl peptide hydrolase.

Formula: C₇H₁₃NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00064441
CAS Number: 65-82-7
EINECS: 200-617-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR