Ac-Lys-nhme

Supplier: BACHEM AMERICAS INC

4000925.0250 4000925.1000
E-1130.0250BAEA 284.04 CAD
E-1130.0250BA E-1130.1000BA
Ac-Lys-nhme
Ac-Lys-nhme

Sequence: Ac-Lys-NHMe


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₉H₁₉N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00044694
CAS Number: 6367-10-8
EINECS: 228-860-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR