Ac-Lys-NH₂·HCl
Supplier: BACHEM AMERICAS INC
Synonyms:
Ac-Lys-NH₂·HCl
4006108.0050
4006108.0250
4006108.1000
E-1125.0050BAEA
185.46
CAD
E-1125.0050BA
E-1125.0250BA
E-1125.1000BA
Ac-Lys-NH₂·HCl
Ac-Lys-NH₂·HCl
Sequence: Ac-Lys-NH₂ · HCl
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₈H₁₈ClN₃O₂ MW: 223.701 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00190718 CAS Number: 104584-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...