AC-Leu-NH₂

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Acetamido-4-methylpentanamide

4000614.0005 4000614.0025
E-1105.0005BAEA 714.25 CAD
E-1105.0005BA E-1105.0025BA
AC-Leu-NH₂
AC-Leu-NH₂

Sequence: Ac-Leu-NH₂


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).


The N-acetylamino acid amides NALA (E-1105), NALMA (E-1110), NAKA (E-1125), NAQA (E-2865), and NATA (E-1225) have been used as model compounds of varying polarity in hydration studies.

Formula: C₈H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00026506
CAS Number: 28529-34-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR