Ac-Ile-NH₂

Supplier: BACHEM AMERICAS INC

4014223.0001 4014223.0005
E-1085.0001BAEA 180.91 CAD
E-1085.0001BA E-1085.0005BA
Ac-Ile-NH₂
Ac-Ile-NH₂

Sequence: Ac-Ile-NH₂

Formula: C₈H₁₆N₂O₂
MW: 172.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080832
CAS Number: 56711-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR