1H-Imidazole-4-propanamide, α(acetylamino)-N-methyl-(S)-

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Acetamido-3-(1H-imidazol-4-yl)-N-methylpropanamide, Ac-His-NHMe

4017306.0250 4017306.1000
E-1070.0250BAEA 1593.94 CAD
E-1070.0250BA E-1070.1000BA
1H-Imidazole-4-propanamide, α(acetylamino)-N-methyl-(S)-
1H-Imidazole-4-propanamide, alpha(acetylamino)-N-methyl-(S)-

Sequence: Ac-His-NHMe


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₉H₁₄N₄O₂
MW: 210.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237489
CAS Number: 6367-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR