p-Nitrophenyl-N-acetylglycinate

Supplier: BACHEM AMERICAS INC

4000250.0001 4000250.0005
E-1065.0001BAEA 180.91 CAD
E-1065.0001BA E-1065.0005BA
p-Nitrophenyl-N-acetylglycinate
p-Nitrophenyl-N-acetylglycinate

Sequence: Ac-Gly-ONp


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
See also Boc-Gly-ONp (A-1740) and Z-Gly-ONp (M-1250).

Formula: C₁₀H₁₀N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00127756
CAS Number: 3304-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR