Ac-asp-OMe

Supplier: BACHEM AMERICAS INC

4000063.0250 4000063.1000
E-1035.0250BAEA 309.81 CAD
E-1035.0250BA E-1035.1000BA
Ac-asp-OMe
Ac-asp-OMe

Sequence: Ac-Asp-OMe


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₇H₁₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD00080825
CAS Number: 4910-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR