AC-Arg-OMe·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Arg-OMe·HCl

4000615.0001 4000615.0005
E-1030.0001BAEA 232.35 CAD
E-1030.0001BA E-1030.0005BA
AC-Arg-OMe·HCl
AC-Arg-OMe·HCl

Sequence: Ac-Arg-OMe · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Substrate for pancreatic and intestinal kallikreins from different species. It is also hydrolyzed by trypsin.

Formula: C₉H₁₉ClN₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00038944
CAS Number: 1784-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR