N-Acetyl-L-arginine

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Arg-OH

4006130.0025 4006130.0100
E-1025.0025BAEA 284.04 CAD
E-1025.0025BA E-1025.0100BA
N-Acetyl-L-arginine
N-Acetyl-L-arginine

Sequence: Ac-Arg-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₈H₁₆N₄O₃
MW: 216.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150285
CAS Number: 155-84-0
EINECS: 205-846-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR