N-Acetyl-L-alanine
Supplier: BACHEM AMERICAS INC
Synonyms:
Ac-Ala-OH, Acetyl-L-alanine
4001766.0025
4001766.0100
4001766.0250
E-1005.0025BAEA
377.3
CAD
E-1005.0025BA
E-1005.0100BA
E-1005.0250BA
N-Acetyl-L-alanine
N-Acetyl-L-alanine
Sequence: Ac-Ala-OH
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
Competitive inhibitor of acyl peptide hydrolase.
Formula:
C₅H₉NO₃ MW: 131.131 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063132 CAS Number: 97-69-8 EINECS: 202-602-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...