4-Methylbenzhydrylamine hydrochloride, polymer supported 100-200 mesh
Supplier: BACHEM AMERICAS INC
4049216.0025
4049216.0100
D-2715.0025BAEA
585.22
CAD
D-2715.0025BA
D-2715.0100BA
4-Methylbenzhydrylamine hydrochloride, polymer supported 100-200 mesh
4-Methylbenzhydrylamine hydrochloride, polymer supported
Sequence: 4-Methyl-benzhydrylamine resin (100-200 mesh) · HCl
Synonym(s): MBHA resin · HCl#4-CH₃-Ph-CH(NH₂)-Ph-polymer · HCl
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...