Fmoc-Lys(biotinyl)-Wang resin 0.3-0.6 mmol/g 200-400 mesh
Supplier: BACHEM AMERICAS INC
4037528.0001
4037528.0005
D-2705.0001BAEA
1497.37
CAD
D-2705.0001BA
D-2705.0005BA
Fmoc-Lys(biotinyl)-Wang resin 0.3-0.6 mmol/g 200-400 mesh
Fmoc-Lys(biotinyl)-Wang resin
Sequence: Fmoc-Lys(biotinyl)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Lys(biotinyl)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...