4-Formyl-3-methoxyphenyloxymethyl on polystyrene resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
Synonyms: 2-Methoxy-4-alkoxybenzaldehyde resin
4029143.0001 4029143.0005
D-2570.0001BAEA 206.51 CAD
D-2570.0001BA D-2570.0005BA
4-Formyl-3-methoxyphenyloxymethyl on polystyrene resin 200-400 mesh
4-Formyl-3-methoxyphenyloxymethyl on polystyrene resin
For Fmoc solid phase synthesis of N-alkyl peptide amides by reductive amination.
Order Now
SPECIFICATIONS
Environmentally Preferable