H-Cys(4-methoxytrityl)-2-chlorotrityl resin 200-400 mesh

Supplier: BACHEM AMERICAS INC

4028399.0001 4028399.0005
D-2495.0001BAEA 903.62 CAD
D-2495.0001BA D-2495.0005BA
H-Cys(4-methoxytrityl)-2-chlorotrityl resin 200-400 mesh
H-Cys(4-methoxytrityl)-2-chlorotrityl resin

Sequence: H-Cys(4-methoxytrityl)-2-chlorotrityl resin (200-400 mesh)
Synonym(s): H-Cys(Mmt)-2-ClTrt resin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR