SASRIN™ resin (100-200 mesh, 0.7-1.0 mmol/g)

Supplier: BACHEM AMERICAS INC

4027899.0001 4027899.0005 4027899.0025
D-2440.0001BAEA 284.04 CAD
D-2440.0001BA D-2440.0005BA D-2440.0025BA
SASRIN™ resin (100-200 mesh, 0.7-1.0 mmol/g)
SASRIN™ resin

SASRIN™ (=Super Acid Sensitive ResIN) was developed in our own laboratories especially for Fmoc solid phase synthesis (EP 0 292 729, US 4,831,084 and US 4,914,151). Its high acid lability enables the cleavage and isolation of the protected peptide fragment.


  • Sequence: 2-Methoxy-4-alkoxybenzyl alcohol resin (200-400 mesh)
  • Substitution: 0.7-1.0 mmol/g


Beside SPPS SASRIN™ has been used in several organic reactions like the synthesis of 4-aryl substituted β-lactams employing Suzuki and Heck couplings, the synthesis of β-sultams, quinazoline-2,4-diones, pyridines and pyrido[2,3-d]pyrimidines and the generation of secondary alkylamines by borane reduction. S.S.Rahman et al. reported on the FT-IR and Raman spectroscopic monitoring of reactions on a single SASRIN™ bead and C.L.Brummel et al. on the evaluation of mass spectrometric methods applicable to the direct analysis of non-peptide bead-bound combinatorial libraries. For a review on the manifold applications of our SASRIN™ resin, including many useful procedures, please ask for our SASRIN™ brochure. Bachem offers a large variety of Fmoc-amino acids linked to SASRIN™ resin. See also D-1295.


For further details see D-1295.

Formula: 3-CH₃O-4-HOCH₂-Ph-O-CH₂-Ph-polymer
Storage Temperature: Refrigerator
MDL Number: MFCD00678022
CAS Number: 124760-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR