Hydroxylamine-Wang resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
Synonyms: 4-H₂N-OCH₂-Ph-O-CH₂-Ph-polymer, Hydroxylamine-p-alkoxybenzyl alcohol resin
4027663.0001 4027663.0005
D-2415.0001BAEA 423.95 CAD
D-2415.0001BA D-2415.0005BA
Hydroxylamine-Wang resin 200-400 mesh
Hydroxylamine-Wang resin
For the synthesis of hydroxamic acids on solid phase. Cleavage from the resin with 70 % TFA in dichloromethane containing 2 % water.
  • Substitution: 0.8-1.1 mmol/g