Fmoc-Tyr(tBu) 0,40-0,90 mmol/g on Wang resin 100-200 mesh

Supplier: BACHEM AMERICAS INC

4028539.0001 4028539.0005
D-2390.0001BAEA 185.46 CAD
D-2390.0001BA D-2390.0005BA
Fmoc-Tyr(tBu) 0,40-0,90 mmol/g on Wang resin 100-200 mesh
Fmoc-Tyr(tBu)

Sequence: Fmoc-Tyr(tBu)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Tyr(tBu)-p-alkoxybenzyl alcohol resin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR