Fmoc-Thr(tBu) 0,40-0,80 mmol/g resin_Wang 100-200 mesh
Supplier: BACHEM AMERICAS INC
4028592.0001
4028592.0005
D-2380.0001BAEA
189.95
CAD
D-2380.0001BA
D-2380.0005BA
Fmoc-Thr(tBu) 0,40-0,80 mmol/g resin_Wang 100-200 mesh
Fmoc-Thr(tBu)
Sequence: Fmoc-Thr(tBu)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Thr(tBu)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...