Fmoc-Ser(tBu) 0.3-0.6 mmol/g on Wang resin 100-200 mesh

Supplier: BACHEM AMERICAS INC

4028572.0001 4028572.0005
D-2375.0001BAEA 180.91 CAD
D-2375.0001BA D-2375.0005BA
Fmoc-Ser(tBu) 0.3-0.6 mmol/g on Wang resin 100-200 mesh
Fmoc-Ser(tBu)

Sequence: Fmoc-Ser(tBu)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Ser(tBu)-p-alkoxybenzyl alcohol resin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR