Fmoc-Lys(boc)-Wang resin 0.5-0.8 mmol/g 100-200 mesh
Supplier: BACHEM AMERICAS INC
4028589.0001
4028589.0005
D-2355.0001BAEA
180.91
CAD
D-2355.0001BA
D-2355.0005BA
Fmoc-Lys(boc)-Wang resin 0.5-0.8 mmol/g 100-200 mesh
Fmoc-Lys(boc)-Wang resin
Sequence: Fmoc-Lys(Boc)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Lys(Boc)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...