Fmoc-Ile 0,50-0,90 mmol/g on Wang resin 100-200 mesh
Supplier: BACHEM AMERICAS INC
4028587.0005
4028587.0025
D-2345.0005BAEA
335.58
CAD
D-2345.0005BA
D-2345.0025BA
Fmoc-Ile 0,50-0,90 mmol/g on Wang resin 100-200 mesh
Fmoc-Ile
Sequence: Fmoc-Ile-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Ile-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...