Fmoc-His(1-Trt)-Wang resin 0.3-0.6 mmol/g 100-200 mesh

Supplier: BACHEM AMERICAS INC

4028586.0001 4028586.0005
D-2340.0001BAEA 232.35 CAD
D-2340.0001BA D-2340.0005BA
Fmoc-His(1-Trt)-Wang resin 0.3-0.6 mmol/g 100-200 mesh
Fmoc-His(1-Trt)-Wang resin

Sequence: Fmoc-His(1-Trt)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-His(1-Trt)-p-alkoxybenzyl alcohol resin#Fmoc-His(τ-Trt)-Wang resin#Fmoc-His(τ-Trt)-p-alkoxybenzyl alcohol resin

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR