Fmoc-Glu(OtBu) 0,5-0,8 mmol/g on Wang resin 100-200 mesh

Supplier: BACHEM AMERICAS INC

4028585.0001 4028585.0005
D-2330.0001BAEA 232.35 CAD
D-2330.0001BA D-2330.0005BA
Fmoc-Glu(OtBu) 0,5-0,8 mmol/g on Wang resin 100-200 mesh
Fmoc-Glu(OtBu)

Sequence: Fmoc-Glu(OtBu)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Glu(OtBu)-p-alkoxybenzyl alcohol resin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR