Fmoc-Gln(Trt) 0.50 - 0.90 mmol/g on Wang resin 100-200 mesh
Supplier: BACHEM AMERICAS INC
4028540.0001
4028540.0005
D-2325.0001BAEA
232.35
CAD
D-2325.0001BA
D-2325.0005BA
Fmoc-Gln(Trt) 0.50 - 0.90 mmol/g on Wang resin 100-200 mesh
Fmoc-Gln(Trt)
Sequence: Fmoc-Gln(Trt)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Gln(Trt)-p-alkoxybenzyl alcohol resin
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...