Fmoc-Cys(Trt) 0.40-1.00 mmol/g on Wang resin 100-200 mesh
Supplier: BACHEM AMERICAS INC
4028211.0001
4028211.0005
D-2320.0001BAEA
232.35
CAD
D-2320.0001BA
D-2320.0005BA
Fmoc-Cys(Trt) 0.40-1.00 mmol/g on Wang resin 100-200 mesh
Fmoc-Cys(Trt)
Sequence: Fmoc-Cys(Trt)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Cys(Trt)-p-alkoxybenzyl alcohol resin
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...