Fmoc-Asn(Trt)-Wang resin 0.40-0.80 mmol/g 100-200 mesh
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-(9H-Fluorene-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanal, Fmoc-L-Asn(Trt)-4-methylbenzhydryl resin, Na-Fmoc-Ng-trityl-L-asparagine 4-alkoxybenzyl alcohol resin
4028584.0001
4028584.0005
D-2310.0001BAEA
232.35
CAD
D-2310.0001BA
D-2310.0005BA
Fmoc-Asn(Trt)-Wang resin 0.40-0.80 mmol/g 100-200 mesh
Fmoc-Asn(Trt)-Wang resin
Sequence: Fmoc-Asn(Trt)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Asn(Trt)-p-alkoxybenzyl alcohol resin
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...