Fmoc-Arg(Pbf) 0.50-1.00 mmol/g on Wang resin 100-200 mesh
Supplier: BACHEM AMERICAS INC
4028394.0001
4028394.0005
D-2305.0001BAEA
309.81
CAD
D-2305.0001BA
D-2305.0005BA
Fmoc-Arg(Pbf) 0.50-1.00 mmol/g on Wang resin 100-200 mesh
Fmoc-Arg(Pbf)
Sequence: Fmoc-Arg(Pbf)-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Arg(Pbf)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...