Fmoc-Ala 0.4-1.0 mmol/g on Wang resin 100-200 mesh
Supplier: BACHEM AMERICAS INC
4027342.0005
4027342.0025
D-2280.0005BAEA
335.58
CAD
D-2280.0005BA
D-2280.0025BA
Fmoc-Ala 0.4-1.0 mmol/g on Wang resin 100-200 mesh
Fmoc-Ala
Sequence: Fmoc-Ala-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Ala-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...