Fmoc-Val 0.50-1.00 mmol/g on Wang resin 100-200 mesh
Supplier: BACHEM AMERICAS INC
4027341.0005
4027341.0025
D-2275.0005BAEA
335.58
CAD
D-2275.0005BA
D-2275.0025BA
Fmoc-Val 0.50-1.00 mmol/g on Wang resin 100-200 mesh
Fmoc-Val
Sequence: Fmoc-Val-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Val-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...