Diphenyldiazomethane resin, polymer-supported 200-400 mesh
Supplier: BACHEM AMERICAS INC
Synonyms:
PDDM resin, Ph-CN₂-Ph-polymer
4027172.0001
4027172.0005
D-2230.0001BAEA
542.13
CAD
D-2230.0001BA
D-2230.0005BA
Diphenyldiazomethane resin, polymer-supported 200-400 mesh
Diphenyldiazomethane resin, polymer-supported
Diphenyldiazomethane was used industrially for producing benzhydryl esters (diphenylmethyl esters) of β-lactam antibiotics. This "polymeric diphenyldiazomethane" readily reacts with carboxylic acids, e.g. Fmoc-amino acids, which need not to be activated. Products can be cleaved from this very acid-sensitive resin derivative with 1-5 % TFA in dichloromethane.
- Substitution: 0.7-1.3 mmol/g