Fmoc-cysteamine-SASRIN™ resin 0.4-0.8 mmol/g 200-400 mesh
Supplier: BACHEM AMERICAS INC
Sequence: Fmoc-cysteamine-SASRIN™ resin (200-400 mesh)
Synonym(s): Fmoc-2-aminoethanethiol-SASRIN™ resin#Fmoc-2-mercaptoethylamine-SASRIN™ resin
Convenient tool for the synthesis of β-thiol containing oligopeptides suitable for chemical conjugation or for the formation of organic monolayers on metal substrates. The cleavage from the resin with TFA/water/triisopropylsilane (94:4:2, v/v/v) yields C-terminal thiols. Linkage of these thiols e.g. to maleimides can be performed under very mild conditions.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...