Chloromethyl polystyrene resin 100-200 mesh

Supplier: BACHEM AMERICAS INC

4031246.0025 4031246.0100
D-2120.0025BAEA 258.14 CAD
D-2120.0025BA D-2120.0100BA
Chloromethyl polystyrene resin 100-200 mesh
Chloromethyl polystyrene resin

  • Substitution: 0.80 - 1.40 mmol/g

Storage Temperature: Ambient
MDL Number: MFCD00146406
CAS Number: 55844-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR