Wang resin 100-200 mesh

Supplier: BACHEM AMERICAS INC

4027344.0025 4027344.0100
D-2115.0025BAEA 619.6 CAD
D-2115.0025BA D-2115.0100BA
Wang resin 100-200 mesh
Wang resin

Substitution: 0.80 - 1.20 mmol/g

Storage Temperature: Ambient
MDL Number: MFCD00131778
CAS Number: 201058-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR