4-(2',4'-Dimethoxyphenyl-fmoc-aminomethyl)phenoxymethyl on polystyrene 0,80-1,20 mmol/g 200-400 mesh
Supplier: BACHEM AMERICAS INC
Synonyms: Rink amide PS resin, 2',4'-Di-CH₃O-Ph-CH(NH-Fmoc)-Ph-4-OCH₂-polystyrene, Rink's resin, Rink amide resin
4026912.0001 4026912.0005 4026912.0025
D-2080.0001BAEA 206.51 CAD
D-2080.0001BA D-2080.0005BA D-2080.0025BA
4-(2',4'-Dimethoxyphenyl-fmoc-aminomethyl)phenoxymethyl on polystyrene 0,80-1,20 mmol/g 200-400 mesh
4-(2',4'-Dimethoxyphenyl-fmoc-aminomethyl)phenoxymethyl on polystyrene
Resin for the Fmoc-SPPS of peptide amides. Cleavage was achieved with 50 % TFA in CH₂Cl₂. or 95% TFA Scavengers may be required. (EP 0 285 562 B1 licensed to Bachem)
  • Substitution: 0.80 - 1.20 mmol/g