Xanthenyl linker resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
Synonyms: (4-(9-Fmoc-amino-xanthen-3-yloxy)-butyryl)-4-methyl-benzhydrylamide resin, Sieber Amide Resin
4026167.0001 4026167.0005 4026167.0025
D-2040.0001BAEA 364.06 CAD
D-2040.0001BA D-2040.0005BA D-2040.0025BA
Xanthenyl linker resin 200-400 mesh
Xanthenyl linker resin
Sieber amide resin has been developed for the Fmoc-SPPS of fully t-butyl protected peptide amides. Cleavage can be achieved with 1% TFA in methylene chloride. Scavengers may be required. As N-methylation increases the acid-lability of the peptide bond, Malakoutikhah et al. employed Sieber resin for obtaining short peptide amides containing multiple MePhe residues.
  • Substitution: 0.4-0.7 mmol/g