H-Cys(trt)-2-chlorotrityl resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
4026170.0001 4026170.0005
D-2015.0001BAEA 903.62 CAD
D-2015.0001BA D-2015.0005BA D-2915.0005BA
H-Cys(trt)-2-chlorotrityl resin 200-400 mesh
H-Cys(trt)-2-chlorotrityl resin
Sequence: H-Cys(Trt)-2-chlorotrityl resin (200-400 mesh)
Peptides containing C-terminal cysteines can be obtained with practically no racemization by Fmoc-based SPPS using 2-chlorotrityl chloride resin.