H-Pro-2-chlorotrityl resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
4025774.0001 4025774.0005
D-2000.0001BAEA 426.05 CAD
D-2000.0001BA D-2000.0005BA
H-Pro-2-chlorotrityl resin 200-400 mesh
H-Pro-2-chlorotrityl resin
Sequence: H-Pro-2-chlorotrityl resin (200-400 mesh)
Peptides containing a C-terminal Pro can be readily obtained on 2-chlorotrityl resin, as diketopiperazine formation is suppressed by the bulky anchor group.