Fmoc-Trp(boc) 0.5-0.8 mmol/g on Wang resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
4025286.0001
4025286.0005
D-1900.0001BAEA
387.21
CAD
D-1900.0001BA
D-1900.0005BA
Fmoc-Trp(boc) 0.5-0.8 mmol/g on Wang resin 200-400 mesh
Fmoc-Trp(boc)
Sequence: Fmoc-Trp(Boc)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Trp(Boc)-p-alkoxybenzyl alcohol resin
Sold under license to US Pat. 5,300,651 and EP 0 516 070
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...