Fmoc-Arg(Pbf) 0.50-1.00 mmol/g on Wang resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
4019093.0001
4019093.0005
D-1860.0001BAEA
370.51
CAD
D-1860.0001BA
D-1860.0005BA
Fmoc-Arg(Pbf) 0.50-1.00 mmol/g on Wang resin 200-400 mesh
Fmoc-Arg(Pbf)
Sequence: Fmoc-Arg(Pbf)-Wang resin (200-400 mesh)
Synonym(s): Fmoc-Arg(Pbf)-p-alkoxybenzyl alcohol resin
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...