Fmoc-Gly 0.50-0.90 mmol/g on Wang resin 100-200 mesh

Supplier: BACHEM AMERICAS INC

4012072.0005 4012072.0025
D-1745.0005BAEA 335.58 CAD
D-1745.0005BA D-1745.0025BA
Fmoc-Gly 0.50-0.90 mmol/g on Wang resin 100-200 mesh
Fmoc-Gly

Sequence: Fmoc-Gly-Wang resin (100-200 mesh)
Synonym(s): Fmoc-Gly-p-alkoxybenzyl alcohol resin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR